Anti-C1R Mouse Monoclonal Antibody [clone: 269104]

Supplier: R & D Systems
MAB1807
220019-370EA 577.02 USD
220019-370
Anti-C1R Mouse Monoclonal Antibody [clone: 269104]
Antibodies
Type: Primary
Antigen: C1R
Clonality: Monoclonal
Clone: 269104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR