Anti-CCR5 Mouse Monoclonal Antibody [clone: CTC5]

Supplier: R & D Systems
MAB1802-100
102885-376EA 633.42 USD
102885-376
Anti-CCR5 Mouse Monoclonal Antibody [clone: CTC5]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: CTC5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR