Anti-HSPD1 Mouse Monoclonal Antibody [clone: 264233]

Supplier: R & D Systems
MAB1800
220019-366EA 600.15 USD
220019-366
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 264233]
Antibodies
Simple Western

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: 264233
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR