Anti-OLR1 Mouse Monoclonal Antibody [clone: 331212]

Supplier: R & D Systems
MAB1798
220019-362EA 647.88 USD
220019-362
Anti-OLR1 Mouse Monoclonal Antibody [clone: 331212]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: 331212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR