Anti-MSR1 Rat Monoclonal Antibody [clone: 268318]

Supplier: R & D Systems
MAB1797
220019-361EA 594.37 USD
220019-361
Anti-MSR1 Rat Monoclonal Antibody [clone: 268318]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 268318
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR