Anti-IL5 Mouse Monoclonal Antibody [clone: 604312]

Supplier: R & D Systems
MAB1795
102885-370EA 772.25 USD
102885-370
Anti-IL5 Mouse Monoclonal Antibody [clone: 604312]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 604312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR