Anti-CCR9 Mouse Monoclonal Antibody [clone: 248621]

Supplier: R & D Systems
MAB1791-100
102885-368EA 616.06 USD
102885-368
Anti-CCR9 Mouse Monoclonal Antibody [clone: 248621]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 248621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR