Anti-CDH11 Mouse Monoclonal Antibody [clone: 667039]

Supplier: R & D Systems
MAB17901
102885-308EA 594.37 USD
102885-308
Anti-CDH11 Mouse Monoclonal Antibody [clone: 667039]
Antibodies
Type: Primary
Antigen: CDH11
Clonality: Monoclonal
Clone: 667039
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR