Anti-CDH11 Mouse Monoclonal Antibody [clone: 283416]

Supplier: R & D Systems
MAB1790
220019-358EA 633.42 USD
220019-358
Anti-CDH11 Mouse Monoclonal Antibody [clone: 283416]
Antibodies
Simple Western

Type: Primary
Antigen: CDH11
Clonality: Monoclonal
Clone: 283416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR