Anti-CCR9 Mouse Monoclonal Antibody [clone: 112509]

Supplier: R & D Systems
MAB179-100
102885-310EA 639.2 USD
102885-310
Anti-CCR9 Mouse Monoclonal Antibody [clone: 112509]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 112509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR