Anti-IFNL3 Rat Monoclonal Antibody [clone: 244710]

Supplier: R & D Systems
MAB1789
220019-355EA 772.25 USD
220019-355
Anti-IFNL3 Rat Monoclonal Antibody [clone: 244710]
Antibodies
Type: Primary
Antigen: IFNL3
Clonality: Monoclonal
Clone: 244710
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR