Anti-LY96 Mouse Monoclonal Antibody [clone: 288307]

Supplier: R & D Systems
MAB1787
220019-354EA 779.48 USD
220019-354
Anti-LY96 Mouse Monoclonal Antibody [clone: 288307]
Antibodies
Type: Primary
Antigen: LY96
Clonality: Monoclonal
Clone: 288307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR