Anti-MMP16 Mouse Monoclonal Antibody [clone: 782005]

Supplier: R & D Systems
MAB1785
102885-306EA 594.37 USD
102885-306
Anti-MMP16 Mouse Monoclonal Antibody [clone: 782005]
Antibodies
Type: Primary
Antigen: MMP16
Clonality: Monoclonal
Clone: 782005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR