Anti-ECE1 Rat Monoclonal Antibody [clone: 303906]

Supplier: R & D Systems
MAB17842
102885-304EA 561.11 USD
102885-304
Anti-ECE1 Rat Monoclonal Antibody [clone: 303906]
Antibodies
Type: Primary
Antigen: ECE1
Clonality: Monoclonal
Clone: 303906
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR