Anti-ECE1 Rat Monoclonal Antibody [clone: 303913]

Supplier: R & D Systems
MAB17841
220019-352EA 561.11 USD
220019-352
Anti-ECE1 Rat Monoclonal Antibody [clone: 303913]
Antibodies
Type: Primary
Antigen: ECE1
Clonality: Monoclonal
Clone: 303913
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR