Anti-ITGB1 Mouse Monoclonal Antibody [clone: 419127]

Supplier: R & D Systems
MAB17783
220019-348EA 600.15 USD
220019-348
Anti-ITGB1 Mouse Monoclonal Antibody [clone: 419127]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: 419127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Pig, Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR