Anti-ITGB1 Mouse Monoclonal Antibody [clone: P5D2]

Supplier: R & D Systems
MAB17781
220019-346EA 655.11 USD
220019-346
Anti-ITGB1 Mouse Monoclonal Antibody [clone: P5D2]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: P5D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR