Anti-ITGB1 Mouse Monoclonal Antibody [clone: 4B7R]

Supplier: R & D Systems
MAB1778
220019-345EA 600.15 USD
220019-345
Anti-ITGB1 Mouse Monoclonal Antibody [clone: 4B7R]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: 4B7R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR