Anti-ITGAX Mouse Monoclonal Antibody [clone: 538207]

Supplier: R & D Systems
MAB17771
220019-344EA 594.37 USD
220019-344
Anti-ITGAX Mouse Monoclonal Antibody [clone: 538207]
Antibodies
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 538207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR