Anti-ITGAX Mouse Monoclonal Antibody [clone: ICRF 3.9]

Supplier: R & D Systems
MAB1777
220019-343EA 763.57 USD
220019-343
Anti-ITGAX Mouse Monoclonal Antibody [clone: ICRF 3.9]
Antibodies
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: ICRF 3.9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR