Anti-MAPK8 Mouse Monoclonal Antibody [clone: 228601]

Supplier: R & D Systems
MAB17761
220019-342EA 594.37 USD
220019-342
Anti-MAPK8 Mouse Monoclonal Antibody [clone: 228601]
Antibodies
Type: Primary
Antigen: MAPK8
Clonality: Monoclonal
Clone: 228601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR