Anti-MAPK9 Mouse Monoclonal Antibody [clone: 228611]

Supplier: R & D Systems
MAB1776
220019-341EA 577.02 USD
220019-341
Anti-MAPK9 Mouse Monoclonal Antibody [clone: 228611]
Antibodies
Type: Primary
Antigen: MAPK9
Clonality: Monoclonal
Clone: 228611
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR