Anti-AKT1 Mouse Monoclonal Antibody [clone: 3247]

Supplier: R & D Systems
MAB17751
220019-339EA 566.89 USD
220019-339
Anti-AKT1 Mouse Monoclonal Antibody [clone: 3247]
Antibodies
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 3247
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR