Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e]

Supplier: R & D Systems
MAB1774
220019-337EA 779.48 USD
220019-337
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e]
Antibodies
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR