Anti-IL17RA Mouse Monoclonal Antibody [clone: 133621]

Supplier: R & D Systems
MAB1771
220019-336EA 763.57 USD
220019-336
Anti-IL17RA Mouse Monoclonal Antibody [clone: 133621]
Antibodies
Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 133621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR