Anti-DKK1 Rat Monoclonal Antibody [clone: 259631]

Supplier: R & D Systems
MAB1765
220019-334EA 594.37 USD
220019-334
Anti-DKK1 Rat Monoclonal Antibody [clone: 259631]
Antibodies
Type: Primary
Antigen: DKK1
Clonality: Monoclonal
Clone: 259631
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR