Anti-POU5F1 Mouse Monoclonal Antibody [clone: 653108]

Supplier: R & D Systems
MAB17591
102885-302EA 647.88 USD
102885-302
Anti-POU5F1 Mouse Monoclonal Antibody [clone: 653108]
Antibodies
Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: 653108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR