Anti-POU5F1 Rat Monoclonal Antibody [clone: 240408]

Supplier: R & D Systems
MAB1759
220019-333EA 647.88 USD
220019-333
Anti-POU5F1 Rat Monoclonal Antibody [clone: 240408]
Antibodies
Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: 240408
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR