Anti-RAET1A (alpha/beta/delta/epsilon) Rat Monoclonal Antibody [clone: 199205]

Supplier: R & D Systems
MAB17581
220019-331EA 763.57 USD
220019-331
Anti-RAET1A (alpha/beta/delta/epsilon) Rat Monoclonal Antibody [clone: 199205]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: RAET1A (alpha/beta/delta/epsilon)
Clonality: Monoclonal
Clone: 199205
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR