Anti-CLEC7A Rat Monoclonal Antibody [clone: 218820]

Supplier: R & D Systems
MAB17561
220019-327EA 594.37 USD
220019-327
Anti-CLEC7A Rat Monoclonal Antibody [clone: 218820]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 218820
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR