Anti-NTRK1 Mouse Monoclonal Antibody [clone: 165126]

Supplier: R & D Systems
MAB175
220019-321EA 763.57 USD
220019-321
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 165126]
Antibodies
Type: Primary
Antigen: NTRK1
Clonality: Monoclonal
Clone: 165126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR