Anti-ROBO1 Mouse Monoclonal Antibody [clone: 243706]

Supplier: R & D Systems
MAB17491
220019-320EA 772.25 USD
220019-320
Anti-ROBO1 Mouse Monoclonal Antibody [clone: 243706]
Antibodies
Type: Primary
Antigen: ROBO1
Clonality: Monoclonal
Clone: 243706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR