Anti-ROBO1 Mouse Monoclonal Antibody [clone: 243716]

Supplier: R & D Systems
MAB1749
220019-319EA 763.57 USD
220019-319
Anti-ROBO1 Mouse Monoclonal Antibody [clone: 243716]
Antibodies
Type: Primary
Antigen: ROBO1
Clonality: Monoclonal
Clone: 243716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR