Anti-TRP53 Rat Monoclonal Antibody [clone: 197643]

Supplier: R & D Systems
MAB1746
220019-316EA 594.37 USD
220019-316
Anti-TRP53 Rat Monoclonal Antibody [clone: 197643]
Antibodies
Type: Primary
Antigen: TRP53
Clonality: Monoclonal
Clone: 197643
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR