Anti-REG3A Mouse Monoclonal Antibody [clone: 250802]

Supplier: R & D Systems
MAB1745
220019-315EA 763.57 USD
220019-315
Anti-REG3A Mouse Monoclonal Antibody [clone: 250802]
Antibodies
Type: Primary
Antigen: REG3A
Clonality: Monoclonal
Clone: 250802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR