Anti-CRP Mouse Monoclonal Antibody [clone: 233539]

Supplier: R & D Systems
MAB17441
220019-314EA 772.25 USD
220019-314
Anti-CRP Mouse Monoclonal Antibody [clone: 233539]
Antibodies
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: 233539
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR