Anti-CRP Mouse Monoclonal Antibody [clone: 233502]

Supplier: R & D Systems
MAB1744
220019-313EA 763.57 USD
220019-313
Anti-CRP Mouse Monoclonal Antibody [clone: 233502]
Antibodies
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: 233502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR