Anti-ITGA2 Rat Monoclonal Antibody [clone: 235033]

Supplier: R & D Systems
MAB1740
220019-311EA 584.25 USD
220019-311
Anti-ITGA2 Rat Monoclonal Antibody [clone: 235033]
Antibodies
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone: 235033
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR