Anti-FSTL1 Rat Monoclonal Antibody [clone: 228208]

Supplier: R & D Systems
MAB17381
220019-308EA 594.37 USD
220019-308
Anti-FSTL1 Rat Monoclonal Antibody [clone: 228208]
Antibodies
Type: Primary
Antigen Symbol: FSTL1
Antigen Name: follistatin-like 1
Antigen Synonyms: Frp, Fstl
Clonality: Monoclonal
Clone: 228208
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR