Anti-AMH Mouse Monoclonal Antibody [clone: 85531]

Supplier: R & D Systems
MAB17371
102885-300EA 577.02 USD
102885-300
Anti-AMH Mouse Monoclonal Antibody [clone: 85531]
Antibodies
Type: Primary
Antigen: AMH
Clonality: Monoclonal
Clone: 85531
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR