Anti-IL22RA2 Rat Monoclonal Antibody [clone: 227702]

Supplier: R & D Systems
MAB1736
220019-305EA 763.57 USD
220019-305
Anti-IL22RA2 Rat Monoclonal Antibody [clone: 227702]
Antibodies
Type: Primary
Antigen: IL22RA2
Clonality: Monoclonal
Clone: 227702
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR