Anti-RECK Mouse Monoclonal Antibody [clone: 198212]

Supplier: R & D Systems
MAB1734
220019-304EA 763.57 USD
220019-304
Anti-RECK Mouse Monoclonal Antibody [clone: 198212]
Antibodies
Type: Primary
Antigen: RECK
Clonality: Monoclonal
Clone: 198212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR