Anti-ITGB2 Mouse Monoclonal Antibody [clone: 212701]

Supplier: R & D Systems
MAB1730
220019-303EA 772.25 USD
220019-303
Anti-ITGB2 Mouse Monoclonal Antibody [clone: 212701]
Antibodies
Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: 212701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR