Anti-SEMA3C Rat Monoclonal Antibody [clone: 238835]

Supplier: R & D Systems
MAB1728
220019-299EA 779.48 USD
220019-299
Anti-SEMA3C Rat Monoclonal Antibody [clone: 238835]
Antibodies
Type: Primary
Antigen: SEMA3C
Clonality: Monoclonal
Clone: 238835
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR