Anti-IL16 Rat Monoclonal Antibody [clone: 228109]

Supplier: R & D Systems
MAB1727
220019-298EA 763.57 USD
220019-298
Anti-IL16 Rat Monoclonal Antibody [clone: 228109]
Antibodies
Type: Primary
Antigen: IL16
Clonality: Monoclonal
Clone: 228109
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR