Anti-FETUB Mouse Monoclonal Antibody [clone: 212621]

Supplier: R & D Systems
MAB1725
220019-296EA 763.57 USD
220019-296
Anti-FETUB Mouse Monoclonal Antibody [clone: 212621]
Antibodies
Type: Primary
Antigen Symbol: FETUB
Antigen Name: fetuin B
Antigen Synonyms: 16G2, Gugu, IRL685
Clonality: Monoclonal
Clone: 212621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR