Anti-MASP1 Mouse Monoclonal Antibody [clone: 254818]

Supplier: R & D Systems
MAB1724
220019-295EA 772.25 USD
220019-295
Anti-MASP1 Mouse Monoclonal Antibody [clone: 254818]
Antibodies
Type: Primary
Antigen: MASP1
Clonality: Monoclonal
Clone: 254818
Conjugation: Unconjugated
Epitope: Catalytic Domain
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR