Anti-DSG3 Mouse Monoclonal Antibody [clone: 216519]

Supplier: R & D Systems
MAB1720
220019-294EA 763.57 USD
220019-294
Anti-DSG3 Mouse Monoclonal Antibody [clone: 216519]
Antibodies
Type: Primary
Antigen: DSG3
Clonality: Monoclonal
Clone: 216519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR