Anti-CXCR4 Mouse Monoclonal Antibody [clone: 44716]

Supplier: R & D Systems
MAB172-100
102885-296EA 655.11 USD
102885-296
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 44716]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 44716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR