Anti-KLK8 Rat Monoclonal Antibody [clone: 369910]

Supplier: R & D Systems
MAB17191
220019-292EA 561.11 USD
220019-292
Anti-KLK8 Rat Monoclonal Antibody [clone: 369910]
Antibodies
Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 369910
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR